Assay Details
| Gene Symbol: LOC4332596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777067.2 | XP_015632553.1 | 1 - 2 | 435 | 74 | |
| GenBank mRNA | AB863631.1 | - | 1 - 2 | 322 | 74 | |
| AK070638.1 | - | 1 - 2 | 408 | 74 | ||
More Information
Set Membership: |
|

LOC4332596
Genomic Map