Assay Details
| Gene Symbol: LOC4332596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777067.2 | XP_015632553.1 | 2 - 3 | 701 | 110 | |
| GenBank mRNA | AB863631.1 | - | 2 - 3 | 588 | 110 | |
| AB863632.1 | - | 2 - 3 | 1284 | 110 | ||
| AK070638.1 | - | 2 - 3 | 674 | 110 | ||
More Information
Set Membership: |
|

LOC4332596
Genomic Map