Assay Details
| Gene Symbol: LOC4332760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777328.2 | XP_015632814.1 | 4 - 5 | 1364 | 67 | |
| GenBank mRNA | AK105307.1 | - | 3 - 4 | 526 | 67 | |
More Information
Set Membership: |
|

LOC4332760
Genomic Map