Assay Details
| Gene Symbol: LOC4333729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772905.2 | XP_015628391.1 | 7 - 8 | 766 | 93 | |
| XM_015772906.2 | XP_015628392.1 | 8 - 9 | 863 | 93 | ||
| GenBank mRNA | AK111767.1 | - | 7 - 8 | 775 | 93 | |
More Information
Set Membership: |
|

LOC4333729
Genomic Map