Assay Details
| Gene Symbol: LOC4333729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772905.2 | XP_015628391.1 | 9 - 10 | 944 | 92 | |
| XM_015772906.2 | XP_015628392.1 | 10 - 11 | 1041 | 92 | ||
| GenBank mRNA | AK111767.1 | - | 9 - 10 | 953 | 92 | |
More Information
Set Membership: |
|

LOC4333729
Genomic Map