Assay Details
| Gene Symbol: LOC4333823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776646.2 | XP_015632132.1 | 5 - 6 | 1462 | 76 | |
| GenBank mRNA | AK072689.1 | - | 5 - 6 | 1463 | 76 | |
| AK072869.1 | - | 6 - 7 | 1134 | 76 | ||
| FJ660625.1 | - | 1 - 2 | 66 | 76 | ||
More Information
Set Membership: |
|

LOC4333823
Genomic Map