Assay Details
| Gene Symbol: LOC4333823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776646.2 | XP_015632132.1 | 6 - 7 | 1568 | 91 | |
| GenBank mRNA | AK071121.1 | - | 1 - 2 | 140 | 91 | |
| AK072689.1 | - | 6 - 7 | 1569 | 91 | ||
| AK072869.1 | - | 7 - 8 | 1240 | 91 | ||
| DQ389164.1 | - | 6 - 7 | 1562 | 91 | ||
| FJ660625.1 | - | 2 - 3 | 172 | 91 | ||
More Information
Set Membership: |
|

LOC4333823
Genomic Map