Assay Details
| Gene Symbol: LOC4333823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776646.2 | XP_015632132.1 | 8 - 9 | 2145 | 74 | |
| XM_015776648.2 | XP_015632134.1 | 7 - 8 | 2031 | 74 | ||
| GenBank mRNA | AK071121.1 | - | 3 - 4 | 717 | 74 | |
| AK072689.1 | - | 8 - 9 | 2146 | 74 | ||
| AK072869.1 | - | 9 - 10 | 1817 | 74 | ||
| AK105792.1 | - | 2 - 3 | 552 | 74 | ||
More Information
Set Membership: |
|

LOC4333823
Genomic Map