Assay Details
| Gene Symbol: LOC4333927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772610.2 | XP_015628096.1 | 3 - 4 | 981 | 67 | |
| GenBank mRNA | AK120822.1 | - | 3 - 4 | 606 | 67 | |
More Information
Set Membership: |
|

LOC4333927
Genomic Map