Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773332.2 | XP_015628818.1 | 3 - 4 | 285 | 62 | |
| XM_015773333.2 | XP_015628819.1 | 2 - 3 | 370 | 62 | ||
| XM_015773334.2 | XP_015628820.1 | 2 - 3 | 369 | 62 | ||
| GenBank mRNA | AK064412.1 | - | 2 - 3 | 167 | 62 | |
| AK106475.1 | - | 2 - 3 | 171 | 62 | ||
| AK108518.1 | - | 2 - 3 | 319 | 62 | ||
| BT131003.1 | - | 2 - 3 | 166 | 62 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map