Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773332.2 | XP_015628818.1 | 7 - 8 | 742 | 99 | |
| XM_015773333.2 | XP_015628819.1 | 7 - 8 | 731 | 99 | ||
| XM_015773334.2 | XP_015628820.1 | 7 - 8 | 730 | 99 | ||
| GenBank mRNA | AK064412.1 | - | 7 - 8 | 528 | 99 | |
| AK106475.1 | - | 6 - 7 | 628 | 99 | ||
| BT131003.1 | - | 7 - 8 | 436 | 99 | ||
| BT132039.1 | - | 3 - 4 | 154 | 99 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map