Assay Details
| Gene Symbol: LOC9269293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795512.2 | XP_015650998.1 | 4 - 5 | 916 | 68 | |
| GenBank mRNA | AK072608.1 | - | 4 - 5 | 861 | 68 | |
| AK121257.1 | - | 3 - 4 | 1136 | 68 | ||
More Information
Set Membership: |
|

LOC9269293
Genomic Map