Assay Details
Gene Symbol: LOC4352218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764854.2 | XP_015620340.1 | 9 - 10 | 1543 | 89 | |
GenBank mRNA | AK073151.1 | - | 9 - 10 | 1531 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
