Assay Details
| Gene Symbol: LOC4330318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767363.1 | XP_015622849.1 | 7 - 8 | 836 | 94 | |
| GenBank mRNA | AK060498.1 | - | 4 - 5 | 375 | 94 | |
| AK102210.1 | - | 7 - 8 | 740 | 94 | ||
More Information
Set Membership: |
|

LOC4330318
Genomic Map