Assay Details
Gene Symbol: LOC4330445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767842.2 | XP_015623328.1 | 7 - 8 | 1387 | 90 | |
GenBank mRNA | AK060937.1 | - | 1 - 2 | 279 | 90 | |
AK102549.1 | - | 7 - 8 | 1220 | 90 | ||
AK102962.1 | - | 5 - 6 | 1504 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
