Assay Details
Gene Symbol: LOC4330445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767842.2 | XP_015623328.1 | 8 - 9 | 1739 | 85 | |
GenBank mRNA | AK060937.1 | - | 2 - 3 | 631 | 85 | |
AK102549.1 | - | 8 - 9 | 1572 | 85 | ||
AK102962.1 | - | 6 - 7 | 1856 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
