Assay Details
| Gene Symbol: LOC4330457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769651.2 | XP_015625137.1 | 1 - 2 | 148 | 135 | |
| GenBank mRNA | AK061019.1 | - | 1 - 2 | 126 | 135 | |
| AK070562.1 | - | 1 - 2 | 101 | 135 | ||
| BT132121.1 | - | 1 - 2 | 121 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769651.2 transcripts.
Set Membership: |
|

LOC4330457
Genomic Map