Assay Details
Gene Symbol: LOC4331056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771351.2 | 1 - 2 | 195 | 53 | ||
GenBank mRNA | AK060165.1 | - | 1 - 2 | 66 | 53 | |
AK067584.1 | - | 1 - 2 | 188 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
