Assay Details
Gene Symbol: LOC4331056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771351.2 | 3 - 4 | 360 | 78 | ||
GenBank mRNA | AK060165.1 | - | 3 - 4 | 231 | 78 | |
AK067584.1 | - | 3 - 4 | 353 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
