Assay Details
Gene Symbol: LOC4331056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771351.2 | 7 - 8 | 666 | 63 | ||
GenBank mRNA | AK060165.1 | - | 7 - 8 | 537 | 63 | |
AK067584.1 | - | 7 - 8 | 659 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
