Assay Details
Gene Symbol: LOC4331869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775122.2 | XP_015630608.1 | 5 - 6 | 603 | 109 | |
GenBank mRNA | AK107127.1 | - | 5 - 6 | 563 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
