Assay Details
| Gene Symbol: LOC4331986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773227.2 | XP_015628713.1 | 1 - 2 | 1616 | 64 | |
| GenBank mRNA | AK121042.1 | - | 1 - 2 | 1583 | 64 | |
More Information
Set Membership: |
|

LOC4331986
Genomic Map