Assay Details
| Gene Symbol: LOC4331986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773227.2 | XP_015628713.1 | 2 - 3 | 1819 | 60 | |
| GenBank mRNA | AK121042.1 | - | 2 - 3 | 1786 | 60 | |
More Information
Set Membership: |
|

LOC4331986
Genomic Map