Assay Details
| Gene Symbol: LOC4332038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773129.2 | XP_015628615.1 | 1 - 2 | 214 | 101 | |
| GenBank mRNA | AK064327.1 | - | 1 - 2 | 200 | 101 | |
| AK102778.1 | - | 1 - 2 | 201 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773129.2 transcripts.
Set Membership: |
|

LOC4332038
Genomic Map