Assay Details
| Gene Symbol: LOC4332038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773129.2 | XP_015628615.1 | 4 - 5 | 801 | 57 | |
| XM_015773130.2 | XP_015628616.1 | 4 - 5 | 795 | 57 | ||
| GenBank mRNA | AK064327.1 | - | 4 - 5 | 787 | 57 | |
| AK102778.1 | - | 4 - 5 | 788 | 57 | ||
More Information
Set Membership: |
|

LOC4332038
Genomic Map