Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 31 - 32 | 4347 | 135 | |
| GenBank mRNA | AK099757.1 | - | 10 - 11 | 1242 | 135 | |
| AK120639.1 | - | 7 - 8 | 761 | 135 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map