Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 36 - 37 | 4877 | 79 | |
| GenBank mRNA | AK099757.1 | - | 15 - 16 | 1772 | 79 | |
| AK120639.1 | - | 12 - 13 | 1302 | 79 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map