Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 39 - 40 | 5281 | 118 | |
| GenBank mRNA | AK099757.1 | - | 18 - 19 | 2176 | 118 | |
| AK120639.1 | - | 15 - 16 | 1706 | 118 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map