Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 41 - 42 | 5500 | 130 | |
| GenBank mRNA | AK099757.1 | - | 20 - 21 | 2395 | 130 | |
| AK120639.1 | - | 17 - 18 | 1925 | 130 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map