Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 46 - 47 | 6156 | 101 | |
| GenBank mRNA | AK099757.1 | - | 25 - 26 | 3051 | 101 | |
| AK120639.1 | - | 22 - 23 | 2581 | 101 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map