Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 24 - 25 | 3558 | 121 | |
| GenBank mRNA | AK099757.1 | - | 3 - 4 | 453 | 121 | |
More Information
Set Membership: |
|

LOC4332045
Genomic Map