Assay Details
Gene Symbol: LOC4332391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775077.2 | XP_015630563.1 | 1 - 2 | 290 | 86 | |
XM_015775078.1 | XP_015630564.1 | 1 - 2 | 218 | 86 | ||
XM_015775079.2 | XP_015630565.1 | 1 - 2 | 218 | 86 | ||
GenBank mRNA | AK066433.1 | - | 1 - 2 | 219 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
