Assay Details
Gene Symbol: LOC4332391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775077.2 | XP_015630563.1 | 2 - 3 | 437 | 66 | |
XM_015775078.1 | XP_015630564.1 | 2 - 3 | 365 | 66 | ||
XM_015775079.2 | XP_015630565.1 | 2 - 3 | 365 | 66 | ||
GenBank mRNA | AK066433.1 | - | 2 - 3 | 366 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
