Assay Details
Gene Symbol: LOC4332391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775077.2 | XP_015630563.1 | 4 - 5 | 706 | 73 | |
GenBank mRNA | AK066433.1 | - | 4 - 5 | 635 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
