Assay Details
Gene Symbol: LOC4332391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775077.2 | XP_015630563.1 | 5 - 6 | 812 | 114 | |
GenBank mRNA | AK066433.1 | - | 5 - 6 | 741 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
