Assay Details
| Gene Symbol: LOC4332477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775503.2 | XP_015630989.1 | 11 - 12 | 1363 | 78 | |
| GenBank mRNA | AK066644.1 | - | 11 - 12 | 1369 | 78 | |
| AK105065.1 | - | 8 - 9 | 721 | 78 | ||
More Information
Set Membership: |
|

LOC4332477
Genomic Map