Assay Details
| Gene Symbol: LOC4332477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775503.2 | XP_015630989.1 | 5 - 6 | 861 | 107 | |
| GenBank mRNA | AK066644.1 | - | 5 - 6 | 867 | 107 | |
| AK105065.1 | - | 2 - 3 | 219 | 107 | ||
More Information
Set Membership: |
|

LOC4332477
Genomic Map