Assay Details
| Gene Symbol: LOC4332979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775971.2 | XP_015631457.1 | 3 - 4 | 608 | 68 | |
| GenBank mRNA | AK068105.1 | - | 3 - 4 | 615 | 68 | |
More Information
Set Membership: |
|

LOC4332979
Genomic Map