Assay Details
Gene Symbol: LOC4333502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776669.2 | XP_015632155.1 | 1 - 2 | 493 | 74 | |
GenBank mRNA | AK111916.1 | - | 1 - 2 | 403 | 74 | |
AK112092.1 | - | 1 - 2 | 484 | 74 | ||
DQ201197.1 | - | 1 - 2 | 125 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
