Assay Details
| Gene Symbol: LOC4333571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776175.2 | XP_015631661.1 | 24 - 25 | 3741 | 75 | |
| GenBank mRNA | AK111443.1 | - | 7 - 8 | 1669 | 75 | |
More Information
Set Membership: |
|

LOC4333571
Genomic Map