Assay Details
| Gene Symbol: LOC4333612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776119.1 | XP_015631605.1 | 1 - 2 | 2989 | 99 | |
| GenBank mRNA | AK073458.1 | - | 1 - 2 | 380 | 99 | |
| AK120058.1 | - | 1 - 2 | 2849 | 99 | ||
| AK120778.1 | - | 1 - 2 | 2998 | 99 | ||
| BT130682.1 | - | 1 - 2 | 355 | 99 | ||
More Information
Set Membership: |
|

LOC4333612
Genomic Map