Assay Details
| Gene Symbol: LOC4334126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776502.2 | XP_015631988.1 | 11 - 12 | 1774 | 92 | |
| GenBank mRNA | AK066914.1 | - | 8 - 9 | 1063 | 92 | |
| AK072597.1 | - | 11 - 12 | 1749 | 92 | ||
More Information
Set Membership: |
|

LOC4334126
Genomic Map