Assay Details
| Gene Symbol: LOC4334126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776502.2 | XP_015631988.1 | 3 - 4 | 630 | 70 | |
| GenBank mRNA | AK061386.1 | - | 3 - 4 | 528 | 70 | |
| AK072597.1 | - | 3 - 4 | 614 | 70 | ||
More Information
Set Membership: |
|

LOC4334126
Genomic Map