Assay Details
| Gene Symbol: LOC4334126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776502.2 | XP_015631988.1 | 9 - 10 | 1245 | 149 | |
| GenBank mRNA | AK066914.1 | - | 6 - 7 | 534 | 149 | |
| AK072597.1 | - | 9 - 10 | 1220 | 149 | ||
More Information
Set Membership: |
|

LOC4334126
Genomic Map