Assay Details
| Gene Symbol: LOC4335126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778384.2 | XP_015633870.1 | 1 | 847 | 67 | |
| GenBank mRNA | AK067184.1 | - | 1 | 853 | 67 | |
More Information
Set Membership: |
|

LOC4335126
Genomic Map