Assay Details
Gene Symbol: LOC4336284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781067.2 | XP_015636553.1 | 1 - 2 | 623 | 69 | |
GenBank mRNA | AK243593.1 | - | 1 - 2 | 497 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
