Assay Details
Gene Symbol: LOC4336284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781067.2 | XP_015636553.1 | 2 - 3 | 931 | 61 | |
GenBank mRNA | AK243593.1 | - | 2 - 3 | 805 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
