Assay Details
| Gene Symbol: LOC4336292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778339.2 | XP_015633825.1 | 3 - 4 | 310 | 68 | |
| GenBank mRNA | AB047923.1 | - | 3 - 4 | 303 | 68 | |
| AK111257.1 | - | 3 - 4 | 302 | 68 | ||
More Information
Set Membership: |
|

LOC4336292
Genomic Map