Assay Details
| Gene Symbol: LOC4336661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781270.2 | XP_015636756.1 | 3 - 4 | 438 | 89 | |
| XM_015781271.2 | XP_015636757.1 | 3 - 4 | 438 | 89 | ||
| XM_015781272.2 | XP_015636758.1 | 3 - 4 | 438 | 89 | ||
| XM_015781273.2 | XP_015636759.1 | 3 - 4 | 438 | 89 | ||
| XM_015781274.2 | XP_015636760.1 | 3 - 4 | 438 | 89 | ||
| GenBank mRNA | AK060894.1 | - | 3 - 4 | 398 | 89 | |
| AK072630.1 | - | 3 - 4 | 444 | 89 | ||
| AK104521.1 | - | 3 - 4 | 398 | 89 | ||
More Information
Set Membership: |
|

LOC4336661
Genomic Map