Assay Details
| Gene Symbol: LOC4336813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780245.2 | XP_015635731.1 | 1 - 2 | 190 | 68 | |
| GenBank mRNA | AK061833.1 | - | 1 - 2 | 183 | 68 | |
| AK098964.1 | - | 1 - 2 | 179 | 68 | ||
| AK100053.1 | - | 1 - 2 | 184 | 68 | ||
More Information
Set Membership: |
|

LOC4336813
Genomic Map